Organic reference standard, 2,3,3',4',6-Pentachlorobiphenyl (PCB No. 110) 35 µg/ml in isooctane, CIL
PROOCIL-PCB-110
:
- Pack type:Glass bottle
- Pk:1 mL
- CAS No.:38380-03-9
- Description:2,3,3',4',6-Pentachlorobiphenyl (PCB No. 110) 35 µg/ml in isooctane
- Formula:C₁₂H₅Cl₅
- Description:2,3,3',4',6-Pentachlorobiphenyl (PCB No. 110)