Organic reference standard, 2,3,3',4',5-Pentachlorobiphenyl (PCB No. 107) 35 µg/ml in isooctane, CIL
Catalog # PROOCIL-PCB-107
Supplier: LGC Standards PROMOCHEM
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:1 mL
- CAS No.:70424-68-9
- Description:2,3,3',4',5-Pentachlorobiphenyl (PCB No. 107) 35 µg/ml in isooctane
- Formula:C₁₂H₅Cl₅
- Description:2,3,3',4',5-Pentachlorobiphenyl (PCB No. 107)