Organic reference standard, 2,3,3',4,4',6-Hexachlorobiphenyl (PCB No. 158) 35 µg/ml in isooctane, CIL
Katalogové číslo PROOCIL-PCB-158
Supplier: LGC Standards PROMOCHEM
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:1 mL
- CAS No.:74472-42-7
- Description:2,3,3',4,4',6-Hexachlorobiphenyl (PCB No. 158) 35 µg/ml in isooctane
- Formula:C₁₂H₄Cl₆
- Description:2,3,3',4,4',6-Hexachlorobiphenyl (PCB No. 158)