Organic reference standard, 2,3',4',5'-Tetrachlorobiphenyl (PCB No. 76) 35 µg/ml in isooctane, CIL
PROOCIL-PCB-76
:
- Pack type:Glass bottle
- Pk:1 mL
- CAS No.:70362-48-0
- Description:2,3',4',5'-Tetrachlorobiphenyl (PCB No. 76) 35 µg/ml in isooctane
- Formula:C₁₂H₆Cl₄
- Description:2,3',4',5'-Tetrachlorobiphenyl (PCB No. 76)