Organic reference standard, 2,2',4-Trichlorobiphenyl (PCB No. 17) 35 µg/ml in isooctane, CIL
Catalog # PROOCIL-PCB-17
Supplier: LGC Standards PROMOCHEM
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:1 mL
- CAS No.:37680-66-3
- Description:2,2',4-Trichlorobiphenyl (PCB No. 17) 35 µg/ml in isooctane
- Formula:C₁₂H₇Cl₃
- Description:2,2',4-Trichlorobiphenyl (PCB No. 17)