Organic reference standard, 2,2',4,5-Tetrachlorobiphenyl (PCB No. 48) 35 µg/ml in isooctane, CIL
Catalog # PROOCIL-PCB-48
Supplier: LGC Standards PROMOCHEM
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:1 mL
- CAS No.:70362-47-9
- Description:2,2',4,5-Tetrachlorobiphenyl (PCB No. 48) 35 µg/ml in isooctane
- Formula:C₁₂H₆Cl₄
- Description:2,2',4,5-Tetrachlorobiphenyl (PCB No. 48)