Organic reference standard, 2,2',3,6-Tetrachlorobiphenyl (PCB No. 45) 35 µg/ml in isooctane, CIL
Catalog # PROOCIL-PCB-45
Dodavatel: LGC Standards PROMOCHEM
Číslo CAS:
Specifikace
- Pack type:Glass bottle
- Pk:1 mL
- CAS No.:70362-45-7
- Description:2,2',3,6-Tetrachlorobiphenyl (PCB No. 45) 35 µg/ml in isooctane
- Formula:C₁₂H₆Cl₄
- Description:2,2',3,6-Tetrachlorobiphenyl (PCB No. 45)