Organic reference standard, 2,2',3,4,6'-Pentachlorobiphenyl (PCB No. 89) 35 µg/ml in isooctane, CIL
Catalog # PROOCIL-PCB-89
Supplier: LGC Standards PROMOCHEM
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:1 mL
- CAS No.:73575-57-2
- Description:2,2',3,4,6'-Pentachlorobiphenyl (PCB No. 89) 35 µg/ml in isooctane
- Formula:C₁₂H₅Cl₅
- Description:2,2',3,4,6'-Pentachlorobiphenyl (PCB No. 89)