Order Entry
Czech Republic
ContactUsLinkComponent
 
Organic reference standard, 2,2',4,5,5'-Pentachlorobiphenyl (PCB No. 101) 100 µg/ml in isooctane, CIL
  PROOCIL-PCB-101-CS
 :  
Organic reference standard, 2,2',4,5,5'-Pentachlorobiphenyl (PCB No. 101) 100 µg/ml in isooctane, CIL
  PROOCIL-PCB-101-CS
 :  

 

  • Pack type:
    Glass bottle
  • Pk:
    1,2 mL
  • CAS No.:
    37680-73-2
  • Description:
    2,2',4,5,5'-Pentachlorobiphenyl (PCB No. 101) 100 µg/ml in isooctane
  • Formula:
    C₁₂H₅Cl₅
  • Description:
    2,2',4,5,5'-Pentachlorobiphenyl (PCB No. 101)