Organic reference standard, 2,2',3,5',6-Pentachlorobiphenyl (PCB No. 95) 35 µg/ml in isooctane, CIL
Katalogové číslo PROOCIL-PCB-95
Dodavatel: LGC Standards PROMOCHEM
Číslo CAS:
Specifikace
- Pack type:Glass bottle
- Pk:1 mL
- CAS No.:38379-99-6
- Description:2,2',3,5',6-Pentachlorobiphenyl (PCB No. 95) 35 µg/ml in isooctane
- Formula:C₁₂H₅Cl₅
- Description:2,2',3,5',6-Pentachlorobiphenyl (PCB No. 95)