Organic reference standard, 2,3',4,4',5'-Pentachlorobiphenyl (PCB No. 123) 100 µg/ml in isooctane, CIL
PROOCIL-PCB-123-CS
:
- Pack type:Glass bottle
- Pk:1,2 mL
- CAS No.:65510-44-3
- Description:2,3',4,4',5'-Pentachlorobiphenyl (PCB No. 123) 100 µg/ml in isooctane
- Formula:C₁₂H₅Cl₅
- Description:2,3',4,4',5'-Pentachlorobiphenyl (PCB No. 123)