Organic reference standard, 2,3,3',4',5'-Pentachlorobiphenyl (PCB No. 122) 35 µg/ml in isooctane, CIL
Catalog # PROOCIL-PCB-122
Supplier: LGC Standards PROMOCHEM
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:1 mL
- CAS No.:76842-07-4
- Description:2,3,3',4',5'-Pentachlorobiphenyl (PCB No. 122) 35 µg/ml in isooctane
- Formula:C₁₂H₅Cl₅
- Description:2,3,3',4',5'-Pentachlorobiphenyl (PCB No. 122)