Organic reference standard, 2,2',3,4',5'-Pentachlorobiphenyl (PCB No. 97) 100 µg/ml in isooctane, CIL
Catalog # PROOCIL-PCB-97-CS
Supplier: LGC Standards PROMOCHEM
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:1,2 mL
- CAS No.:41464-51-1
- Description:2,2',3,4',5'-Pentachlorobiphenyl (PCB No. 97) 100 µg/ml in isooctane
- Formula:C₁₂H₅Cl₅
- Description:2,2',3,4',5'-Pentachlorobiphenyl (PCB No. 97)