Order Entry
Czech Republic
ContactUsLinkComponent
 
Organic reference standard, 2,2',5,6'-Tetrachlorobiphenyl (PCB No. 53) 10 µg/ml in isooctane, Dr Ehrenstorfer
  EHERL20005300IO
 :  
Organic reference standard, 2,2',5,6'-Tetrachlorobiphenyl (PCB No. 53) 10 µg/ml in isooctane, Dr Ehrenstorfer
  EHERL20005300IO
 :  

 

  • Pack type:
    Glass bottle
  • Pk:
    10 mL
  • CAS No.:
    41464-41-9
  • Description:
    2,2',5,6'-Tetrachlorobiphenyl (PCB No. 53) 10 µg/ml in isooctane
  • Formula:
    C₁₂H₆Cl₄
  • Description:
    2,2',5,6'-Tetrachlorobiphenyl (PCB No. 53