Organic reference standard, 2,3',4,5'-Tetrachlorobiphenyl (PCB No. 68) 10 µg/ml in isooctane, Dr Ehrenstorfer
EHERL20006800IO
:
- Pack type:Glass bottle
- Pk:10 mL
- CAS No.:73575-52-7
- Description:2,3',4,5'-Tetrachlorobiphenyl (PCB No. 68) 10 µg/ml in isooctane
- Formula:C₁₂H₆Cl₄
- Description:2,3',4,5'-Tetrachlorobiphenyl (PCB No. 68