Organic reference standard, 2,6-Dichlorobiphenyl (PCB No. 10) 35 µg/ml in isooctane, CIL
Catalog # PROOCIL-PCB-10
Supplier: LGC Standards PROMOCHEM
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:1 mL
- CAS No.:33146-45-1
- Description:2,6-Dichlorobiphenyl (PCB No. 10) 35 µg/ml in isooctane
- Formula:C₁₂H₈Cl₂
- Description:2,6-Dichlorobiphenyl (PCB No. 10)