Order Entry
Czech Republic
ContactUsLinkComponent
 
Organic reference standard, 3,4-Dichlorobiphenyl (PCB No. 12) 35 µg/ml in isooctane, CIL
  PROOCIL-PCB-12
 :  
Organic reference standard, 3,4-Dichlorobiphenyl (PCB No. 12) 35 µg/ml in isooctane, CIL
  PROOCIL-PCB-12
 :  

 

  • Pack type:
    Glass bottle
  • Pk:
    1 mL
  • CAS No.:
    2974-92-7
  • Description:
    3,4-Dichlorobiphenyl (PCB No. 12) 35 µg/ml in isooctane
  • Formula:
    C₁₂H₈Cl₂
  • Description:
    3,4-Dichlorobiphenyl (PCB No. 12)