Organic reference standard, 2,2',3,6'-Tetrachlorobiphenyl (PCB No. 46) 10 µg/ml in isooctane, Dr Ehrenstorfer
EHERL20004600IO
:
- Pack type:Glass bottle
- Pk:10 mL
- CAS No.:41464-47-5
- Description:2,2',3,6'-Tetrachlorobiphenyl (PCB No. 46) 10 µg/ml in isooctane
- Formula:C₁₂H₆Cl₄
- Description:2,2',3,6'-Tetrachlorobiphenyl (PCB No. 46