
L(+)-Arginine (not of animal origin) 98.5-101.0% Ph. Eur., USP cell culture reagent, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:74-79-3
:
Formula:C₆H₁₄N₄O₂
MDL Number:MFCD00002635
:174.2 g/mol
Merck Index:13,00785
Boiling Pt:368 °C (1013 hPa)
Melting Pt:238 °C
Density:1.3 g/cm³ (20 °C)
:M-Clarity™ quality level = MQ200
EINECS:200-811-1
:GHS07
Storage Temperature:Ambient
Specification Test Results
- Appearance (Color):White
- Appearance (Form):Crystalline Powder
- Solubility (Color):Colorless
- Solubility (Turbidity) at 1.0 g to 10 ml of H2O:Clear
- Endotoxin Assay:< 1 EU/mg
- Identity by IR:Conforms to Structure
- Origin (Must Be Non-Animal Source):Non-Animal Source
- Specific Rotation C = 8 in 6 M HCl (20 Deg C/EP):26.9 - 27.9 °
- Specific Rotation C = 8 in 6 M HCl (25 Deg C/USP):26.3 - 27.7 °
- Ammonium:<= 0.02%
- Sulfate:<= 0.020%
- Iron (Fe):<= 10 ppm
- Chloride:<= 0.020%
- Loss on Drying:< 0.50 %
- Residue on ignition (Ash):< 0.10 %
- Ninhydrin-Positive Substances (Other amino acids):Conforms
- pH (5% in H2O):10.5 - 12.0
- Assay (Dry Basis):98.5 - 101.0 %
Specification Test Results
Frequently Bought Together