Order Entry
Czech Republic
ContactUsLinkComponent
Organic reference standard, 2,2',3,4',5',6-Hexachlorobiphenyl (PCB No. 149) 35 µg/ml in isooctane, CIL
Organic reference standard, 2,2',3,4',5',6-Hexachlorobiphenyl (PCB No. 149) 35 µg/ml in isooctane, CIL
Catalog # PROOCIL-PCB-149
CAS Number:  
Organic reference standard, 2,2',3,4',5',6-Hexachlorobiphenyl (PCB No. 149) 35 µg/ml in isooctane, CIL
Catalog # PROOCIL-PCB-149
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1 mL
  • CAS No.:
    38380-04-0
  • Description:
    2,2',3,4',5',6-Hexachlorobiphenyl (PCB No. 149) 35 µg/ml in isooctane
  • Formula:
    C₁₂H₄Cl₆
  • Description:
    2,2',3,4',5',6-Hexachlorobiphenyl (PCB No. 149)

Specifications

About this item