Organic reference standard, 2,2',3,4',5,5'-Hexachlorobiphenyl (PCB No. 146) 10 µg/ml in isooctane, Dr Ehrenstorfer
Catalog # EHERL20014600IO
Supplier: EHRENSTORFER
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:10 mL
- CAS No.:51908-16-8
- Description:2,2',3,4',5,5'-Hexachlorobiphenyl (PCB No. 146) 10 µg/ml in isooctane
- Formula:C₁₂H₄Cl₆
- Description:2,2',3,4',5,5'-Hexachlorobiphenyl (PCB No. 146)