Order Entry
Czech Republic
ContactUsLinkComponent
 
Organic reference standard, 2,3',4,4',5,5'-Hexachlorobiphenyl (PCB No. 167) 35 µg/ml in isooctane, CIL
  PROOCIL-PCB-167
 :  
Organic reference standard, 2,3',4,4',5,5'-Hexachlorobiphenyl (PCB No. 167) 35 µg/ml in isooctane, CIL
  PROOCIL-PCB-167
 :  

 

  • Pack type:
    Glass bottle
  • Pk:
    1 mL
  • CAS No.:
    52663-72-6
  • Description:
    2,3',4,4',5,5'-Hexachlorobiphenyl (PCB No. 167) 35 µg/ml in isooctane
  • Formula:
    C₁₂H₄Cl₆
  • Description:
    2,3',4,4',5,5'-Hexachlorobiphenyl (PCB No. 167)