Organic reference standard, 2,3,3',4',5,5',6-Heptachlorobiphenyl (PCB No. 193) 10 µg/ml in isooctane, Dr Ehrenstorfer
EHERL20019300IO
:
- Pack type:Glass bottle
- Pk:10 mL
- CAS No.:69782-91-8
- Description:2,3,3',4',5,5',6-Heptachlorobiphenyl (PCB No. 193) 10 µg/ml in isooctane
- Formula:C₁₂H₃Cl₇
- Description:2,3,3',4',5,5',6-Heptachlorobiphenyl (PCB No. 193)