Order Entry
Czech Republic
ContactUsLinkComponent
Organic reference standard, 2,2',3,3',4,4',6,6'-Octachlorobiphenyl (PCB No. 197) 35 µg/ml in isooctane, CIL
Organic reference standard, 2,2',3,3',4,4',6,6'-Octachlorobiphenyl (PCB No. 197) 35 µg/ml in isooctane, CIL
  PROOCIL-PCB-197
 :  
Organic reference standard, 2,2',3,3',4,4',6,6'-Octachlorobiphenyl (PCB No. 197) 35 µg/ml in isooctane, CIL
  PROOCIL-PCB-197
 :  

 

  • Pack type:
    Glass bottle
  • Pk:
    1 mL
  • CAS No.:
    33091-17-7
  • Description:
    2,2',3,3',4,4',6,6'-Octachlorobiphenyl (PCB No. 197) 35 µg/ml in isooctane
  • Formula:
    C₁₂H₂Cl₈
  • Description:
    2,2',3,3',4,4',6,6'-Octachlorobiphenyl (PCB No. 197)