Organic reference standard, 2,2',3,3',4,4',5-Heptachlorobiphenyl (PCB No. 170) 100 µg/ml in isooctane, CIL
Catalog # PROOCIL-PCB-170-CS
Supplier: LGC Standards PROMOCHEM
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:1,2 mL
- CAS No.:35065-30-6
- Description:2,2',3,3',4,4',5-Heptachlorobiphenyl (PCB No. 170) 100 µg/ml in isooctane
- Formula:C₁₂H₃Cl₇
- Description:2,2',3,3',4,4',5-Heptachlorobiphenyl (PCB No. 170)