Organic reference standard, 2,2',3,3',4,4',5,5'-Octachlorobiphenyl (PCB No. 194) 10 µg/ml in isooctane, Dr Ehrenstorfer
Catalog # EHERL20019400IO
Supplier: EHRENSTORFER
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:10 mL
- CAS No.:35694-08-7
- Description:2,2',3,3',4,4',5,5'-Octachlorobiphenyl (PCB No. 194) 10 µg/ml in isooctane
- Formula:C₁₂H₂Cl₈
- Description:2,2',3,3',4,4',5,5'-Octachlorobiphenyl (PCB No. 194)