Order Entry
Czech Republic
ContactUsLinkComponent
Organic reference standard, 2,3,4,4',5,6-Hexachlorobiphenyl (PCB No. 166) 35 µg/ml in isooctane, CIL
Organic reference standard, 2,3,4,4',5,6-Hexachlorobiphenyl (PCB No. 166) 35 µg/ml in isooctane, CIL
  PROOCIL-PCB-166
 :  
Organic reference standard, 2,3,4,4',5,6-Hexachlorobiphenyl (PCB No. 166) 35 µg/ml in isooctane, CIL
  PROOCIL-PCB-166
 :  

 

  • Pack type:
    Glass bottle
  • Pk:
    1 mL
  • CAS No.:
    41411-63-6
  • Description:
    2,3,4,4',5,6-Hexachlorobiphenyl (PCB No. 166) 35 µg/ml in isooctane
  • Formula:
    C₁₂H₄Cl₆
  • Description:
    2,3,4,4',5,6-Hexachlorobiphenyl (PCB No. 166)