Organic reference standard, 2,3,3',4,5,5',6-Heptachlorobiphenyl (PCB No. 192) 35 µg/ml in isooctane, CIL
PROOCIL-PCB-192
:
- Pack type:Glass bottle
- Pk:1 mL
- CAS No.:74472-51-8
- Description:2,3,3',4,5,5',6-Heptachlorobiphenyl (PCB No. 192) 35 µg/ml in isooctane
- Formula:C₁₂H₃Cl₇
- Description:2,3,3',4,5,5',6-Heptachlorobiphenyl (PCB No. 192)