VWR®, Organic reference standard, Chloroethane 200 µg/ml in methanol
Catalog # 123482M
Supplier: VWR Chemicals
CAS Number:
Specifications
- Pack type:Glass ampoule
- Pk:1 mL
- CAS No.:75-00-3
- Description:Chloroethane 200 µg/ml in methanol
- Formula:C₂H₅Cl
- Description:Chloroethane