Order Entry
Czech Republic
ContactUsLinkComponent
Supelco® Organic Reference Standard, 1,2-Dibromoethane solution
Supelco® Organic Reference Standard, 1,2-Dibromoethane solution
Catalog # 48880-U
Supplier:  Merck
CAS Number:  
undefined
Supelco® Organic Reference Standard, 1,2-Dibromoethane solution
Catalog # 48880-U
Supplier:  Merck
CAS Number:  

Specifications

  • Concentration:
    200 μg/ml in methanol
  • Pk:
    1
  • Standards:
    Certified reference material
  • CAS No.:
    106-93-4
  • Description:
    1,2-Dibromoethane solution
  • Storage conditions:
    2...30 °C
  • Hazard class description:
    H225 - H301 + H311 + H331 - H370
  • Shipping conditions:
    STD
  • MDL No.:
    MFCD00000233
  • M-Clarity quality level:
    MQ100
  • Synonym(s):
    Ethylene dibromide
  • Molecular weight:
    187,86
  • Flash point:
    11 °C

Specifications

About this item

  • GHS signal: GHS02, GHS06, GHS08
Caution: P210 - P233 - P280 - P301 + P310 - P303 + P361 + P353 - P304 + P340 + P311