Order Entry
Czech Republic
ContactUsLinkComponent
 
Health
Irritant
Environment
Nickel(II) sulphate hexahydrate ≥98%, GPR RECTAPUR®
Catalog # 25895.294
Supplier:  VWR Chemicals
CAS Number:  
undefined
Health
Irritant
Environment
Nickel(II) sulphate hexahydrate ≥98%, GPR RECTAPUR®
Catalog # 25895.294
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle for solids
  • Pk:
    1 kg
  • Seq:
    4024

Specifications

Specification Test Results

  • Assay:
    Min. 98 %
  • Cl (Chloride):
    Max. 0.02 %
  • Cd (Cadmium):
    Max. 25 ppm
  • Cu (Copper):
    Max. 25 ppm
  • Pb (Lead):
    Max. 25 ppm
  • Zn (Zinc):
    Max. 25 ppm

Specification Test Results

About this item

 :
 :262.85 g/mol
Merck Index:13,06541
Melting Pt:840 °C
Density:2.07 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:232-104-9
UN:3288
ADR:6.1,III
 :GHS08
 :GHS07
 :GHS09
CAS Number:10101-97-0
Formula:NiSO₄·6H₂O
MDL Number:MFCD00149813