
Ruthenium(IV) oxide hydrate ≥99.99% (metals basis) Ru 54-60%, Premion®
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:32740-79-7
:
Formula:O₂Ru·H₂O
MDL Number:MFCD00149846
:151.08 g/mol
Melting Pt:1200 °C (subl.)
Density:6.97 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:234-840-6
UN:1479
ADR:,
:GHS07
Related Information
Frequently Bought Together