Order Entry
Czech Republic
ContactUsLinkComponent
Platinium(IV) oxide hydrate hydrogenation catalyst, for synthesis, Sigma-Aldrich®
Oxidizing
Corrosive
Platinium(IV) oxide hydrate hydrogenation catalyst, for synthesis, Sigma-Aldrich®
Catalog # 8.07346.0001
Supplier:  Merck
CAS Number:  
Oxidizing
Corrosive
Platinium(IV) oxide hydrate hydrogenation catalyst, for synthesis, Sigma-Aldrich®
Catalog # 8.07346.0001
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

Specification Test Results

  • Solubility:
    (20 °C) insoluble
  • Melting Point:
    450 °C
  • Molar Mass:
    227.08 g/mol

Specification Test Results

About this item

 :
 :245.09 g/mol
Merck Index:12,07686
Melting Pt:450 °C
Density:10.2 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:215-223-0
UN:3085
ADR:5.1 (8),II
 :M-Clarity™ quality level = MQ200
 :GHS03
 :GHS05
CAS Number:52785-06-5
 :MFCD00011184
Formula:O₂Pt·H₂O
MDL Number:MFCD00066964