Iridium(IV) oxide dihydrate, Sigma-Aldrich®
Catalog # SIAL398500-1G
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:260.25 g/mol
:M-Clarity™ quality level = MQ100
EINECS:234-743-9
CAS Number:30980-84-8
Formula:IrO₂·2H₂O
MDL Number:MFCD00011065