




Cobalt(II) nitrate hexahydrate, EMSURE® for analysis, Supelco®
Supplier: Merck
Certificates
About this item
CAS Number:10026-22-9
:
Formula:Co(NO₃)₂·6H₂O
MDL Number:MFCD00149647
:291.03 g/mol
Merck Index:13,02469
Melting Pt:57 °C
Density:1.88 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:233-402-1
ADR:9,II
:M-Clarity™ quality level = MQ300
UN:3077
:GHS08
:GHS07
:GHS03
:GHS09
:GHS05
Specification Test Results
- Solubility:2170 g/l (100 °C)
- Melting Point:57 °C
- Molar Mass:291.04 g/mol
- Bulk Density:800 kg/m3
- Density:1.87 g/cm3 (20 °C)
- pH:4.0 (100 g/l, H2O, 20 °C)
- Hazard Class:9
- Hazard Class Description:Miscellaneous Hazardous Material
Specification Test Results
Related Information
Frequently Bought Together