


Cobalt(II) nitrate hexahydrate 98.0-102.0% ACS
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:10026-22-9
:
Formula:Co(NO₃)₂·6H₂O
MDL Number:MFCD00149647
:291.03 g/mol
Merck Index:14,2444
Melting Pt:55…56 °C
Density:1.87
EINECS:233-402-1
UN:1477
ADR:5.1,III
:GHS03
:GHS08
:GHS07
Storage Temperature:Ambient
Related Information
Frequently Bought Together