

Ruthenium(III) chloride hydrate, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:14898-67-0
:20759-14-2
Formula:Cl₃Ru·xH₂O
MDL Number:MFCD00149844
:207.43 g/mol (anhy)
Melting Pt:> 300 °C
Density:3.11 g/cm³ (20 °C)
EINECS:233-167-5
UN:3260
ADR:8,III
:GHS05
:GHS07
Frequently Bought Together