

Ruthenium(III) chloride hydrate ≥99.99% (metals basis), Premion®
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:14898-67-0
:20759-14-2
Formula:Cl₃Ru·xH₂O
MDL Number:MFCD00149844
:207.43 g/mol (anhy)
Merck Index:14,8302
Melting Pt:100 °C (dec.)
Density:3.11 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:233-167-5
UN:3260
ADR:8,III
:GHS05
:GHS07
Related Information
Frequently Bought Together