


Nickel(II) chloride hexahydrate, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:7791-20-0
:
Formula:NiCl₂·6H₂O
MDL Number:MFCD00149809
:237.69 g/mol
Merck Index:13,06529
Melting Pt:140 °C
Density:1.84 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:231-743-0
ADR:6.1,III
:M-Clarity™ quality level = MQ200
UN:3288
:GHS06
:GHS08
:GHS09
Frequently Bought Together