


Nickel(II) chloride hexahydrate ≥98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:7791-20-0
:
Formula:NiCl₂·6H₂O
MDL Number:MFCD00149809
:237.69 g/mol
Merck Index:14,6505
Melting Pt:140 °C (-H₂O)
Density:1.92
EINECS:231-743-0
UN:3288
ADR:6.1,III
:GHS06
:GHS08
:GHS09
Storage Temperature:Ambient
Related Information
Frequently Bought Together