Order Entry
Czech Republic
ContactUsLinkComponent
Nickel(II) chloride hexahydrate 99.999% (trace metals basis)
Toxic
Health
Environment
Nickel(II) chloride hexahydrate 99.999% (trace metals basis)
Certificates
undefined
Toxic
Health
Environment
Nickel(II) chloride hexahydrate 99.999% (trace metals basis)

Specification Test Results

  • Appearance:
    Green Crystalline powder or crystals and/or chunks
  • Total trace metal impurities:
    ≤0.001 %
  • Nickel (Ni):
    24 to 26 %

Specification Test Results

Related Information

Frequently Bought Together