

Iron(III) chloride hexahydrate ≥97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:10025-77-1
:
Formula:FeCl₃·6H₂O
MDL Number:MFCD00149712
:270.3 g/mol
Merck Index:14,4019
Boiling Pt:280…285 °C
Melting Pt:37 °C
Density:1.820
Storage Temperature:Ambient
EINECS:231-729-4
UN:3260
ADR:8,III
:GHS05
:GHS07
Frequently Bought Together