ContactUsLinkComponent
Iron(III) chloride hexahydrate 99+%, extra pure
undefined
Iron(III) chloride hexahydrate 99+%, extra pure
  • Appearance:
    Yellow to brown Granules or lumps
  • Titration Iodometric:
    ≥99.0 %
  • Sulfate (SO4):
    ≤1500 ppm
  • Ferrous iron (Fe 2+):
    ≤0.9 %
  • Acidity (HCl):
    ≤0.8 %
  • Insoluble matter:
    ≤0.05 % (10 g in 50 ml H2O:HCl (49:1))
Frequently Bought Together