

Iron(II) chloride tetrahydrate, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:13478-10-9
:
Formula:FeCl₂·4H₂O
MDL Number:MFCD00149709
Melting Pt:677 °C
Density:1.926 g/cm³ (20 °C)
Storage Temperature:Ambient
ADR:8,III
:M-Clarity™ quality level = MQ200
:198.81 g/mol
EINECS:231-843-4
UN:3260
:GHS05
:GHS07
Frequently Bought Together