


Cobalt(II) chloride hexahydrate, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:7791-13-1
:
Formula:CoCl₂·6H₂O
MDL Number:MFCD00149652
:237.93 g/mol
Merck Index:13,02462
Boiling Pt:110 °C (1013 hPa)
Melting Pt:56 °C
Density:1.92 g/cm³ (20 °C)
Storage Temperature:Ambient
UN:3077
ADR:9,III
:M-Clarity™ quality level = MQ200
EINECS:231-589-4
:GHS08
:GHS07
:GHS09
Frequently Bought Together