Order Entry
Czech Republic
ContactUsLinkComponent
 
Health
Environment
Manganese(II) sulphate monohydrate, BAKER ANALYZED® ACS, J.T.Baker®
Catalog # 0174.1000
Supplier:  Avantor
CAS Number:  
Health
Environment
Manganese(II) sulphate monohydrate, BAKER ANALYZED® ACS, J.T.Baker®
Catalog # 0174.1000
Supplier:  Avantor
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle for solids
  • Pk:
    1 kg
  • Seq:
    4024

Specifications

Specification Test Results

  • Assay:
    98.0 - 101.0 %
  • Calcium (Ca):
    max. 0.005 %
  • Chloride (Cl):
    max. 0.001 %
  • Iron (Fe):
    max. 0.001 %
  • Potassium (K):
    max. 0.005 %
  • Magnesium (Mg):
    max. 0.005 %
  • Sodium (Na):
    max. 0.005 %
  • Zinc (Zn):
    max. 0.005 %
  • Heavy Metals (as Pb):
    max. 0.002 %
  • Insoluble Matter:
    max. 0.01 %
  • Loss on Ignition:
    10.0 - 12.0 %
  • Substances Reducing KMnO4:
    passes test
  • Copper (Cu):
    max. 5
  • Nickel (Ni):
    max. 5

Specification Test Results

About this item

 :
Merck Index:13,05763
Melting Pt:700 °C
Density:2.95 g/cm³ (25 °C)
ADR:9,III
 :169.02 g/mol
EINECS:232-089-9
UN:3077
 :GHS08
 :GHS09
Storage Temperature:Refrigerator
CAS Number:10034-96-5
Formula:MnSO₄·1H₂O
MDL Number:MFCD00149159