Order Entry
Czech Republic
ContactUsLinkComponent
Manganese(II) sulphate monohydrate 99+%, extra pure
Corrosive
Health
Environment
Manganese(II) sulphate monohydrate 99+%, extra pure
Certificates
undefined
Corrosive
Health
Environment
Manganese(II) sulphate monohydrate 99+%, extra pure
Certificates

Specification Test Results

  • Appearance:
    White to light pink Powder
  • Titration Complexometric:
    99.0 to 101.0 %
  • Loss on ignition:
    10.0 % to 12.0 % (500°C)
  • Calcium (Ca):
    ≤200 ppm
  • Iron (Fe):
    ≤20 ppm
  • Magnesium (Mg):
    ≤200 ppm
  • Zinc (Zn):
    ≤50 ppm
  • KMnO4-reducing substance:
    Passes test

Specification Test Results

Related Information

Frequently Bought Together