
Manganese(II) nitrate hexahydrate ≥98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:17141-63-8
:
Formula:Mn(NO₃)₂·6H₂O
:287.04 g/mol
Merck Index:13,05756
Melting Pt:24.9 °C
Density:1.72 g/cm³ (30 °C)
Storage Temperature:Ambient
EINECS:233-828-8
UN:2724
ADR:5.1,III
:GHS03
Related Information
Frequently Bought Together