



Manganese(II) nitrate tetrahydrate, EMSURE® for analysis, Supelco®
: Merck
:20694-39-7
:
:Mn(NO₃)₂·4H₂O
:MFCD00149789
:251.01 g/mol
:37 °C
:2.115 g/cm³ (20 °C)
:Ambient
:233-828-8
:2724
:5.1,III
:M-Clarity™ quality level = MQ300
:GHS03
:GHS05
:GHS08
:GHS07
- Solubility:3800 g/l (20 °C)
- Melting Point:37 °C
- Molar Mass:251.01 g/mol
- Bulk Density:750 - 950 kg/m3
- Density:2.13 g/cm3 (20 °C)
- pH:3 (50 g/l, H2O, 20 °C)
- Hazard Class:5.1
- Hazard Class Description:Oxidizer
Frequently Bought Together