



Manganese(II) nitrate tetrahydrate ≥98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:20694-39-7
:
Formula:Mn(NO₃)₂·4H₂O
MDL Number:MFCD00149789
:251.01 g/mol
Merck Index:14,5733
Melting Pt:∼26 °C
Density:2.115 g/cm³ (20 °C)
EINECS:233-828-8
UN:2724
ADR:5.1,III
:GHS03
:GHS05
:GHS08
:GHS07
Storage Temperature:Ambient
Frequently Bought Together